Merck
  • Interdependence of threonine, methionine and isoleucine metabolism in plants: accumulation and transcriptional regulation under abiotic stress.
Interdependence of threonine, methionine and isoleucine metabolism in plants: accumulation and transcriptional regulation under abiotic stress.
Amino acids (2010-02-27)
Vijay Joshi, Je-Gun Joung, Zhangjun Fei, Georg Jander
ABSTRACT
Pathways regulating threonine, methionine and isoleucine metabolism are very efficiently interconnected in plants. As both threonine and methionine serve as substrates for isoleucine synthesis, their synthesis and catabolism under different developmental and environmental conditions also influence isoleucine availability. Together, methionine gamma-lyase and threonine deaminase maintain the isoleucine equilibrium in plants under varied substrate availabilities. Isoleucine and the two other branched-chain amino acids (BCAAs) (leucine and valine) share four common enzymes in their biosynthesis pathways and thus are coordinately regulated. Induction of free amino acids as osmolytes in response to abiotic stress is thought to play a role in plant stress tolerance. In particular, the accumulation of BCAAs is induced many-fold during osmotic stress. However, unlike in the case of proline, not much research has been focused on understanding the function of the response involving BCAAs. This review describes pathways influencing branched-chain amino acid metabolism and what is known about the biological significance of their accumulation under abiotic stress. A bioinformatics approach to understanding the transcriptional regulation of the genes involved in amino acid metabolism under abiotic stress is also presented.
Product Description
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Isoleucine
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Isoleucine, European Pharmacopoeia (EP) Reference Standard