Merck
Proline metabolism and microenvironmental stress.
Annual review of nutrition (2010-04-27)
James M Phang, Wei Liu, Olga Zabirnyk
ABSTRACT
Proline, the only proteinogenic secondary amino acid, is metabolized by its own family of enzymes responding to metabolic stress and participating in metabolic signaling. Collagen in extracellular matrix, connective tissue, and bone is an abundant reservoir for proline. Matrix metalloproteinases degrading collagen are activated during stress to make proline available, and proline oxidase, the first enzyme in proline degradation, is induced by p53, peroxisome proliferator-activated receptor gamma (PPARgamma) and its ligands, and by AMP-activated protein kinase downregulating mTOR. Metabolism of proline generates electrons to produce ROS and initiates a variety of downstream effects, including blockade of the cell cycle, autophagy, and apoptosis. The electrons can also enter the electron transport chain to produce adenosine triphosphate for survival under nutrient stress. Pyrroline-5-carboxylate, the product of proline oxidation, is recycled back to proline with redox transfers or is sequentially converted to glutamate and alpha-ketoglutarate. The latter augments the prolyl hydroxylation of hypoxia-inducible factor-1alpha and its proteasomal degradation. These effects of proline oxidase, as well as its decreased levels in tumors, support its role as a tumor suppressor. The mechanism for its decrease is mediated by a specific microRNA. The metabolic signaling by proline oxidase between oxidized low-density lipoproteins and autophagy provides a functional link between obesity and increased cancer risk.
Product Description
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Proline, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Proline
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material