Merck
  • Muscle cramps in cirrhosis: the therapeutic value of quinine. Is it underused?
Muscle cramps in cirrhosis: the therapeutic value of quinine. Is it underused?
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver (2008-03-25)
A Corbani, P Manousou, V Calvaruso, I Xirouchakis, A K Burroughs
ABSTRACT
Muscle cramps are a common and recurring symptom in patients with cirrhosis. Although, the pathophysiology has not been specifically studied in cirrhosis, this is thought to be the same for cramps in general, originating in the motorneurone, with high frequency firing of motor unit action potentials. However precise pathophysiological mechanisms are not known. Risk factors in cirrhosis have been little studied. Neither aetiology, nor pre-ascitic or ascitic stage, nor electrolyte disturbances, nor use of diuretic therapy has been found to have a statistical association with cramps in patients with cirrhosis. Effective treatments, from this literature review, are albumin, which however is expensive and has little applicability as preventative therapy and oral quinine or quinidine. Quinine is little used in Italy but licensed in the UK for the therapy of muscle cramps. There is evidence for the efficacy of quinine in patients without cirrhosis and in healthy subjects. In cirrhosis quinidine (isomer of quinine) has also been shown to be effective versus placebo. Its major effect is in the prevention of cramps. More widespread use of quinine and further studies are needed, particularly in Italy and other countries, in which its use has been limited, as it is effective therapy in many patients with cirrhosis.
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Supelco
Quinine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Quinine, 90%