Merck
  • Perspectives of biotechnological production of L-tyrosine and its applications.
Perspectives of biotechnological production of L-tyrosine and its applications.
Applied microbiology and biotechnology (2007-10-31)
Tina Lütke-Eversloh, Christine Nicole S Santos, Gregory Stephanopoulos
ABSTRACT
The aromatic amino acid L-tyrosine is used as a dietary supplement and has promise as a valuable precursor compound for various industrial and pharmaceutical applications. In contrast to chemical production, biotechnological methods can produce L-tyrosine from biomass feedstocks under environmentally friendly and near carbon-free conditions. In this minireview, various strategies for synthesizing L-tyrosine by employing biocatalysts are discussed, including initial approaches as well as more recent advances. Whereas early attempts to engineer L-tyrosine-excreting microbes were based on auxotrophic and antimetabolite-resistant mutants, recombinant deoxyribonucleic acid technology and a vastly increasing knowledge of bacterial physiology allowed recently for more targeted genetic manipulations and strain improvements. As an alternative route, L-tyrosine can also be obtained from the conversion of phenol, pyruvate, and ammonia or phenol and serine in reactions catalyzed by the enzyme tyrosine phenol lyase.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, FG
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Tyrosine, European Pharmacopoeia (EP) Reference Standard