Merck
Direct and indirect cellular effects of aspartame on the brain.
European journal of clinical nutrition (2007-08-09)
P Humphries, E Pretorius, H Naudé
ABSTRACT
The use of the artificial sweetener, aspartame, has long been contemplated and studied by various researchers, and people are concerned about its negative effects. Aspartame is composed of phenylalanine (50%), aspartic acid (40%) and methanol (10%). Phenylalanine plays an important role in neurotransmitter regulation, whereas aspartic acid is also thought to play a role as an excitatory neurotransmitter in the central nervous system. Glutamate, asparagines and glutamine are formed from their precursor, aspartic acid. Methanol, which forms 10% of the broken down product, is converted in the body to formate, which can either be excreted or can give rise to formaldehyde, diketopiperazine (a carcinogen) and a number of other highly toxic derivatives. Previously, it has been reported that consumption of aspartame could cause neurological and behavioural disturbances in sensitive individuals. Headaches, insomnia and seizures are also some of the neurological effects that have been encountered, and these may be accredited to changes in regional brain concentrations of catecholamines, which include norepinephrine, epinephrine and dopamine. The aim of this study was to discuss the direct and indirect cellular effects of aspartame on the brain, and we propose that excessive aspartame ingestion might be involved in the pathogenesis of certain mental disorders (DSM-IV-TR 2000) and also in compromised learning and emotional functioning.
Product Description
Supelco
Aspartame, analytical standard
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%
USP
Aspartame, United States Pharmacopeia (USP) Reference Standard
Aspartame, European Pharmacopoeia (EP) Reference Standard
Supelco
Aspartame, Pharmaceutical Secondary Standard; Certified Reference Material