Merck
Lanthanum carbonate as a first-line phosphate binder: the "cons".
Seminars in dialysis (2007-07-20)
Tilman B Drüeke
ABSTRACT
Controlling serum phosphorus levels in patients with renal failure is critical. The use of oral phosphate-binding agents is universal for patients with end-stage kidney disease to reduce phosphate absorption. The therapeutic goal is to reduce serum phosphorus levels without disturbing calcium homeostasis or promoting accumulation of potentially toxic elements from the medication. Aluminum hydroxide effectively reduces serum phosphorus, but has largely been abandoned as a first-line phosphate binder because of hazards associated with metal absorption and tissue accumulation. Traditional calcium-based phosphate binders tend to promote hypercalcemia and calcium overloading, and are linked to accelerated cardiovascular calcification. Interest in aluminum-free, calcium-free phosphate-binding agents continues to grow. Sevelamer hydrochloride, a metal-free, calcium-free hydrogel, is not absorbed, has been proven safe and efficacious in controlling serum phosphorus, and is associated with attenuated progression of cardiovascular calcification. Lanthanum carbonate is a newer aluminum-free, calcium-free phosphate-binding agent. Lanthanum is a rare-earth trace metal with industrial and agricultural applications. As a therapeutic, this metal-based binder appears effective in reducing serum phosphorus, yet concerns remain about lanthanum accumulation in tissues during long-term oral administration. Similar to the metal aluminum, lanthanum is absorbed in the intestine and accumulates in body tissues, especially in the liver, bone, muscle, kidney, and brain. Moreover, the rate of intestinal absorption of lanthanum is enhanced in chronic renal failure. Our experience with aluminum hydroxide suggests caution regarding the long-term use of another metal-based agent that displays enhanced absorption in the uremic state and progressive tissue accumulation.
Sigma-Aldrich
Lanthanum, pieces
Lanthanum, rod, 100mm, diameter 6.35mm, cast, 99%
Lanthanum, rod, 50mm, diameter 6.35mm, cast, 99%
Lanthanum, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Lanthanum, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, 100x100mm, thickness 0.1mm, as rolled, 99%
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Lanthanum, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Lanthanum, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, 100x100mm, thickness 0.125mm, as rolled, 99%