Merck
  • A review of workplace aerosol sampling procedures and their relevance to the assessment of beryllium exposures.
A review of workplace aerosol sampling procedures and their relevance to the assessment of beryllium exposures.
Journal of environmental monitoring : JEM (2006-06-13)
Martin Harper
ABSTRACT
Standardized conventions governing the fractions of airborne particles that can penetrate the human head airways, the thoracic airways and the alveolar spaces have been internationally (although not universally) adopted. Several agencies involved in setting limit values for occupational exposure concentrations have taken these conventions into account when considering the appropriate standard for specific chemicals, in order to ensure the standards are biologically relevant. A convention is selected based on the characteristic health effects, and forms the basis of measurement against the limiting concentration value. In order to assess exposure for comparison to this metric or any other purposes, it is necessary to choose a sampler whose performance matches the convention, and protocols have been developed and used to test sampler performance. Several aerosol sampling devices are available, nominally at least, for each of the conventions. Some considerations important to the sampling of airborne particles containing beryllium with regard to the sampling conventions, the test protocols and sampler performance are discussed.
Product Description
Beryllium, wire reel, 1m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 1m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.3m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.85mm, annealed and clean, 99%