Merck
Update on the use of ciclosporin in immune-mediated dermatoses.
The British journal of dermatology (2006-06-16)
C E M Griffiths, A Katsambas, B A C Dijkmans, A Y Finlay, V C Ho, A Johnston, T A Luger, U Mrowietz, K Thestrup-Pedersen
ABSTRACT
Immune-mediated dermatoses, such as psoriasis and atopic dermatitis, affect a significant proportion of the population. Although most cases are not life threatening, these diseases can have a profound effect on the sufferer's quality of life and that of their family. Systemic therapy, such as ciclosporin, is often indicated for severe or recalcitrant disease. The efficacy of ciclosporin in the treatment of psoriasis and atopic dermatitis has been established and clinical data also demonstrate its efficacy in treating less common but equally challenging conditions such as pyoderma gangrenosum, lichen planus, autoimmune bullous disease, recalcitrant chronic idiopathic urticaria and chronic dermatitis of the hands and feet. The risk of potential adverse events associated with ciclosporin is greatly reduced if current treatment and monitoring guidelines are followed.
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyclosporin A solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Ciclosporin, European Pharmacopoeia (EP) Reference Standard