Merck
  • Analysis of five HMG-CoA reductase inhibitors-- atorvastatin, lovastatin, pravastatin, rosuvastatin and simvastatin: pharmacological, pharmacokinetic and analytical overview and development of a new method for use in pharmaceutical formulations analysis and in vitro metabolism studies.
Analysis of five HMG-CoA reductase inhibitors-- atorvastatin, lovastatin, pravastatin, rosuvastatin and simvastatin: pharmacological, pharmacokinetic and analytical overview and development of a new method for use in pharmaceutical formulations analysis and in vitro metabolism studies.
Biomedical chromatography : BMC (2005-09-07)
Md Khalid Pasha, Syed Muzeeb, Shaik Jafar Sadik Basha, Dhanya Shashikumar, Ramesh Mullangi, Nuggehally R Srinivas
ABSTRACT
A specific, accurate, precise and reproducible high-performance liquid chromatographic (HPLC) method was developed and validated for the simultaneous quantitation of five 3-hydroxy-3-methyglutaryl coenzyme A (HMG-CoA) reductase inhibitors, viz. atorvastatin, lovastatin, pravastatin, rosuvastatin and simvastatin, in pharmaceutical formulations and extended the application to in vitro metabolism studies of these statins. Ternary gradient elution at a flow rate of 1 mL/min was employed on an Intertisl ODS 3V column (4.6 x 250 mm, 5 microm) at ambient temperature. The mobile phase consisted of 0.01 m ammonium acetate (pH 5.0), acetonitrile and methanol. Theophylline was used as an internal standard (IS). The HMG-CoA reductase inhibitors and their metabolites were monitored at a wavelength of 237 nm. Drugs were found to be 89.6-105.6% of their label's claim in the pharmaceutical formulations. For in vitro metabolism studies the reaction mixtures were extracted with simple liquid-liquid extraction using ethyl acetate. Baseline separation of statins and their metabolites along with IS free from endogenous interferences was achieved. Nominal retention times of IS, atorvastatin, lovastatin, pravastatin, rosuvastatin and simvastatin were 7.5, 17.2, 21.6, 28.5, 33.5 and 35.5 min, respectively. The proposed method is simple, selective and could be applicable for routine analysis of HMG-CoA reductase inhibitors in pharmaceutical preparations as well as in vitro metabolism studies.
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Lovastatin for peak identification, European Pharmacopoeia (EP) Reference Standard