Merck
  • Polymers derived from the amino acid L-tyrosine: polycarbonates, polyarylates and copolymers with poly(ethylene glycol).
Polymers derived from the amino acid L-tyrosine: polycarbonates, polyarylates and copolymers with poly(ethylene glycol).
Advanced drug delivery reviews (2003-04-23)
Sharon L Bourke, Joachim Kohn
ABSTRACT
The natural amino acid L-tyrosine is a major nutrient having a phenolic hydroxyl group. This feature makes it possible to use derivatives of tyrosine dipeptide as a motif to generate diphenolic monomers, which are important building blocks for the design of biodegradable polymers. Particularly useful monomers are desaminotyrosyl-tyrosine alkyl esters (abbreviated as DTR, where R stands for the specific alkyl ester used). Using this approach, a wide variety of polymers have been synthesized. Here, tyrosine-derived polycarbonates, polyarylates, and polyethers are reviewed with special emphasis on recent developments relating to cellular and in vivo responses, sterilization techniques, surface characterization, drug delivery, and processing and fabrication techniques. The commercial development of tyrosine-derived polycarbonates is most advanced, with one polymer, poly(DTE carbonate) (E=ethyl), being under review by the USA Federal Drug Administration.
Product Description
Tyrosine, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard