Merck
  • Alfacalcidol and calcitriol in the prevention of bone loss after organ transplantation.
Alfacalcidol and calcitriol in the prevention of bone loss after organ transplantation.
Calcified tissue international (1999-09-15)
P Sambrook
ABSTRACT
There is increasing evidence that D-hormones (active vitamin D metabolites) are effective agents in preventing posttransplantation bone loss of various organs. This effect may relate to several mechanisms including reversal of secondary hyperparathyroidism from cyclosporine, reversal of corticosteroid effects on bone, or immunomodulatory effects with corticosteroid sparing. Further clinical trials are necessary to establish their comparative efficacy to other agents, but D-hormones should be considered as prophylactic therapy in patients undergoing organ transplantation.
Sigma-Aldrich
1α-Hydroxyvitamin D3, ≥97.0% (HPLC)
Alfacalcidol, European Pharmacopoeia (EP) Reference Standard