Merck
  • Impairment of renal structure and function following heterogeneous chemical mixture exposure in rats.
Impairment of renal structure and function following heterogeneous chemical mixture exposure in rats.
Indian journal of experimental biology (2014-04-30)
Kiran Morya, Kauresh D Vachhrajani
ABSTRACT
Renal structural and functional alterations following an exposure to a heterogeneous chemical mixture (HCM) of phthalic acid di butyl ester, 1, 2-dichlorobenzene, cadmium chloride and chromium trioxide, administered through oral gavage in low doses (1/100 and 1/1000 of LD50 value of individual chemical) for 60 days, followed by withdrawal till 120 days resulted in significant rise in kidney lipid peroxidation and fall in the activities of enzymatic antioxidants. However, withdrawal of HCM treatment restored most of these altered parameters. Degenerative changes in the kidney included proximal convoluted tubules devoid of brush boarder with cytoplasmic blebbing, dissolution and sloughing of nuclei. Cortical glomeruli were also affected with epithelial disintegration, pyknosis of podocyte nuclei and mesengial cell hyperplasia. The morphological alterations recovered fully in the low dose compared to the high dose treatment group.
Product Description
Supelco
1,2-Dichlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Chromium(VI) oxide, 99.99% trace metals basis
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Phthalic acid, analytical standard
Sigma-Aldrich
Chromium(VI) oxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
1,2-Dichlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Dichlorobenzene, suitable for HPLC, 99%
Sigma-Aldrich
1,2-Dichlorobenzene solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cadmium chloride, technical grade
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)