Merck
  • Integrative refolding and purification of histidine-tagged protein by like-charge facilitated refolding and metal-chelate affinity adsorption.
Integrative refolding and purification of histidine-tagged protein by like-charge facilitated refolding and metal-chelate affinity adsorption.
Journal of chromatography. A (2014-04-29)
Hu Liu, Wen-Jie Du, Xiao-Yan Dong, Yan Sun
ABSTRACT
This work proposed an integrative method of protein refolding and purification by like-charged resin facilitated refolding and metal-chelate affinity adsorption. Hexahistidine-tagged enhanced green fluorescence protein (EGFP) was overexpressed in Escherichia coli as inclusion bodies (IBs), and then the protein was refolded and purified from urea-solubilized IBs by this method. A metal-chelating resin was fabricated by coupling iminodiacetic acid (IDA) to agarose gel (Sepharose FF). The anionic resin was used to facilitate the refolding of like-charged EGFP from IBs. After refolding, nickel ions were introduced for the affinity purification of the target protein by metal-chelating adsorption. It was found that the resin was effective in facilitating EGFP refolding. For 0.1mg/mL EGFP IBs refolding, the fluorescence recovery (FR) by direct dilution was only 64%; addition of only 0.05 g/mL resin increased the FR to over 90%. Moreover, the FR increased with increasing resin concentration. Owning to the shielding effect of the oppositely charged impurities embedded in IBs on the surface charges of the IDA resin, more resin particles were required to exert an aggregation inhibition effect in the IBs protein refolding. Additionally, compared with direct-dilution refolding, inclusion of like-charged resins not only offered an enhanced FR of EGFP, but also bound some opposite-charged contaminant proteins, leading to a preliminary purification effect. Afterwards, the refolded EGFP was recovered by metal-chelating adsorption at an FR of 85% and purity of 93%. This work has thus extended the like-charge facilitated protein refolding strategy to the integrative protein refolding and purification.
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, Type I, low EEO
SAFC
L-Histidine
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%