Merck
  • Adsorption of methyl violet onto mesoporous MCM-48 from aqueous solution.
Adsorption of methyl violet onto mesoporous MCM-48 from aqueous solution.
Journal of nanoscience and nanotechnology (2014-04-18)
Xingxing Gu, Hong Xu, Lingling Luo, Jun Wu, Hongjun Lin, Jianrong Chen
ABSTRACT
In this study, hexadecyltrimethyl ammonium bromide and triblock poly(ethylene oxide)-poly(propylene oxide)-poly(ethylene oxide) were used as co-templates and tetraethoxysilane was used as silica source to synthesize mesoporous MCM-48, which was employed to adsorb methyl violet dye from water. The prepared MCM-48, after calcination at 550 degrees C, was found to have a high surface area of 1072 m2/g and a pore volume of 1.08 cm3/g. The MCM-48 adsorption of methyl violet in aqueous solution was studied using UV-visible spectrophotometry. Experimental conditions, including initial pH of sample solution, initial concentration, MCM-48 amount, adsorption time and temperature, were also investigated. Results showed that the adsorption behavior could well be depicted by Langmuir equations and pseudo-second-order kinetic model. The maximum adsorption capacity of 193.82 mg/g was obtained at 20 degrees C. The values for thermodynamic parameters deltaG0, deltaS0 and deltaH0 were all negative, showing that the MCM-48 adsorption of methyl violet was spontaneous and exothermic.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet
Sigma-Aldrich
Crystal Violet, certified by the BSC
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Zeolite
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Aluminosilicate, mesostructured, MCM-41 (hexagonal)
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard