Merck
  • Differential toxicity of rod and spherical zinc oxide nanoparticles on human peripheral blood mononuclear cells.
Differential toxicity of rod and spherical zinc oxide nanoparticles on human peripheral blood mononuclear cells.
Journal of biomedical nanotechnology (2014-04-17)
Debalina Bhattacharya, Chitta Ranjan Santra, Amar N Ghosh, Parimal Karmakar
ABSTRACT
The cytotoxic and genotoxic potential of rod and spherical zinc oxide nanoparticles (ZnO NPs) were evaluated on human peripheral blood mononuclear cells (PBMCs). Their sizes and shapes were determined by Transmission electron microscopy (TEM) and physical characterizations were done by X-ray diffraction (XRD) and Dynamic light scattering (DLS). The cytotoxic potential of rod ZnO NPs was greater than spherical ZnO NPs when applied to PBMCs and was limited to the proliferative lymphocytes. Rod ZnO NPs produced more reactive oxygen species (ROS) compared to spherical ones. Additionally rod ZnO NPs induced significant DNA damage to PBMCs as revealed by cytokinesis block micronucleus (CBMN) assay and comet assay. On the other hand UV absorption property was enhanced in case of spherical ZnO NPs compared to rod ZnO NPs. Thus, present study implicates the shape dependent differential application of ZnO NPs in our daily life.
Product Description
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, nanowires, Length 1 um
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%