Merck
Case report, aetiology, and treatment of an acquired long-QT syndrome.
Acta clinica Belgica (2014-04-15)
P J Van Asbroeck, W Huybrechts, R De Soir
ABSTRACT
Acquired long-QT syndrome is an iatrogenic disorder, usually induced by drugs, which can cause life-threatening arrhythmias. We present a case report on an acquired long-QT syndrome with an interesting confluence of circumstances, and comment on aetiology and treatment.
Sigma-Aldrich
(±)-Sotalol hydrochloride, ≥98% (TLC), powder