Merck
  • S-nitrosylation influences the structure and DNA binding activity of AtMYB30 transcription factor from Arabidopsis thaliana.
S-nitrosylation influences the structure and DNA binding activity of AtMYB30 transcription factor from Arabidopsis thaliana.
Biochimica et biophysica acta (2014-03-04)
Carolina Pereira Tavares, Javier Vernal, Ricardo Alexandre Delena, Lorenzo Lamattina, Raul Cassia, Hernán Terenzi
ABSTRACT
MYB proteins are a family of transcription factors that play an important role in plant development and regulatory defense processes. Arabidopsis thaliana MYB30 (AtMYB30), a member of this protein family, is involved in cell death processes during the hypersensitive response (HR) of plants. HR is characterized by a vast production of reactive oxygen species (ROS) and nitric oxide (NO). NO may thus influence the binding of AtMYB30 to DNA. In this work we evaluated the effect of NO on AtMYB30 DNA binding activity, and also in the protein structural properties. A fully active minimal DNA-binding domain (DBD) of AtMYB30 (residues 11-116) containing two cysteine residues (C49 and C53) was overexpressed and purified. Site-directed mutagenesis was used to obtain AtMYB30 DBD mutants C49A and C53A. The DNA binding activity of AtMYB30 DBD, and Cys single mutants is clearly inhibited upon incubation with a NO donor, and S-nitrosylation was confirmed by the biotin switch assay. Finally, in order to understand the mechanism of NO effect on AtMYB30 DNA binding activity we performed circular dichroism analysis, to correlate the observed protein function inhibition and a potential structural impairment on AtMYB30 DBD. Indeed, NO modification of C49 and C53 residues promotes a subtle modification on the secondary structure of this transcription factor. We thus demonstrated, using various techniques, the in vitro effect of NO on AtMYB30 DBD, and thus the potential consequences of NO activity on plant metabolism influenced by this transcription factor.
Product Description
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
SAFC
BIOTIN
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material