Merck
  • Hyperglycemia diverts dividing osteoblastic precursor cells to an adipogenic pathway and induces synthesis of a hyaluronan matrix that is adhesive for monocytes.
Hyperglycemia diverts dividing osteoblastic precursor cells to an adipogenic pathway and induces synthesis of a hyaluronan matrix that is adhesive for monocytes.
The Journal of biological chemistry (2014-02-27)
Aimin Wang, Ronald J Midura, Amit Vasanji, Andrew J Wang, Vincent C Hascall
ABSTRACT
Isolated rat bone marrow stromal cells cultured in osteogenic medium in which the normal 5.6 mm glucose is changed to hyperglycemic 25.6 mm glucose greatly increase lipid formation between 21-31 days of culture that is associated with decreased biomineralization, up-regulate expression of cyclin D3 and two adipogenic markers (CCAAT/enhancer binding protein α and peroxisome proliferator-activated receptor γ) within 5 days of culture, increase neutral and polar lipid synthesis within 5 days of culture, and form a monocyte-adhesive hyaluronan matrix through an endoplasmic reticulum stress-induced autophagic mechanism. Evidence is also provided that, by 4 weeks after diabetes onset in the streptozotocin-induced diabetic rat model, there is a large loss of trabecular bone mineral density without apparent proportional changes in underlying collagen matrices, a large accumulation of a hyaluronan matrix within the trabecular bone marrow, and adipocytes and macrophages embedded in this hyaluronan matrix. These results support the hypothesis that hyperglycemia in bone marrow diverts dividing osteoblastic precursor cells (bone marrow stromal cells) to a metabolically stressed adipogenic pathway that induces synthesis of a hyaluronan matrix that recruits inflammatory cells and establishes a chronic inflammatory process that demineralizes trabecular cancellous bone.
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 500,000-750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,750,000-2,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 130,000-150,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,250,000-1,500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-110,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 150,000-300,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor