Merck
  • Expression and interaction of small heat shock proteins (sHsps) in rice in response to heat stress.
Expression and interaction of small heat shock proteins (sHsps) in rice in response to heat stress.
Biochimica et biophysica acta (2014-02-26)
Xinhai Chen, Shoukai Lin, Qiulin Liu, Jian Huang, Wenfeng Zhang, Jun Lin, Yongfei Wang, Yuqin Ke, Huaqin He
ABSTRACT
The inherent immobility of rice (Oryza sativa L.) limited their abilities to avoid heat stress and required them to contend with heat stress through innate defense abilities in which heat shock proteins played important roles. In this study, Hsp26.7, Hsp23.2, Hsp17.9A, Hsp17.4 and Hsp16.9A were up-regulated in Nipponbare during seedling and anthesis stages in response to heat stress. Subsequently, the expressing levels of these five sHsps in the heat-tolerant rice cultivar, Co39, were all significantly higher than that in the heat-susceptible rice cultivar, Azucena. This indicated that the expressive level of these five sHsps was positively related to the ability of rice plants to avoid heat stress. Thus, the expression level of these five sHsps can be regarded as bio-markers for screening rice cultivars with different abilities to avoid heat stress. Hsp18.1, Hsp17.9A, Hsp17.7 and Hsp16.9A, in the three rice cultivars under heat stress were found to be involved in one protein complex by Native-PAGE, and the interactions of Hsp18.1 and Hsp 17.7, Hsp18.1 and Hsp 17.9A, and Hsp17.7 and Hsp16.9A were further validated by yeast 2-hybridization. Pull down assay also confirmed the interaction between Hsp17.7 and Hsp16.9A in rice under heat stress. In conclusion, the up-regulation of the 5 sHsps is a key step for rice to tolerate heat stress, after that some sHsps assembled into a large hetero-oligomeric complex. In addition, through protein-protein interaction, Hsp101 regulated thiamine biosynthesis, and Hsp82 homology affected nitrogen metabolism, while Hsp81-1 were involved in the maintenance of sugar or starch synthesis in rice plants under heat stress. These results provide new insight into the regulatory mechanism of sHsps in rice.
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch from corn
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice