Merck
  • MED15, encoding a subunit of the mediator complex, is overexpressed at high frequency in castration-resistant prostate cancer.
MED15, encoding a subunit of the mediator complex, is overexpressed at high frequency in castration-resistant prostate cancer.
International journal of cancer (2014-01-01)
Zaki Shaikhibrahim, Roopika Menon, Martin Braun, Anne Offermann, Angela Queisser, Diana Boehm, Wenzel Vogel, Kerstin Rüenauver, Christian Ruiz, Tobias Zellweger, Maria Svensson, Ove Andren, Glen Kristiansen, Nicolas Wernert, Lukas Bubendorf, Jutta Kirfel, Saskia Biskup, Sven Perner
ABSTRACT
The mediator complex is an evolutionary conserved key regulator of transcription of protein-coding genes and an integrative hub for diverse signaling pathways. In this study, we investigated whether the mediator subunit MED15 is implicated in castration-resistant prostate cancer (CRPC). MED15 expression and copy number/rearrangement status were assessed by immunohistochemistry (IHC) and fluorescence in situ hybridization (FISH), respectively on 718 prostate cancer (PCa) specimens and sequenced by Sanger on a subset. Furthermore, SMAD3 phosphorylation, androgen receptor (AR) and proliferation markers were evaluated by IHC. In PCa cells, siRNA/shRNA knockdown of MED15 was followed by proliferation assays with/without dihydrotestosterone (DHT), and treatments with recombinant TGF-β3. Our results show that MED15 is overexpressed in 76% of distant metastatic CRPC (CRPC(MET) ) and 70% of local-recurrent CRPC (CRPC(LOC) ), in contrast to low frequencies in androgen-sensitive PCa, and no expression in benign prostatic tissue. Furthermore, MED15 overexpression correlates with worse clinical outcome thus defining a highly lethal phenotype. Moreover, TGF-β signaling activation associates with MED15 overexpression in PCa tissues, and leads to increased expression of MED15 in PCa cells. MED15 knockdown effects phosphorylation and shuttling of p-SMAD3 to the nucleus as well as TGF-β-enhanced proliferation. In PCa tissues, MED15 overexpression associates with AR overexpression/amplification and correlates with high proliferative activity. MED15 knockdown decreases both androgen-dependent and -independent proliferation in PCa cells. Taken together, these findings implicate MED15 in CRPC, and as MED15 is evolutionary conserved, it is likely to emerge as a lethal phenotype in other therapeutic-resistant diseases, and not restricted to our disease model.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Glycine 1 M solution