Merck
Loa loa-does it deserve to be neglected?
The Lancet. Infectious diseases (2013-12-18)
Wolfram Gottfried Metzger, Benjamin Mordmüller
ABSTRACT
More than 10 million people in western and central Africa are estimated to be infected with Loa loa filarial nematodes. Like most other infectious diseases, L loa filariasis (loiasis) covers a wide range of symptoms. Severe complications have been reported; however, most observations are anecdotal, typically in travellers. The widespread use of filaricidal drugs within eradication programmes of Onchocerca volvulus and Wuchereria bancrofti led to the observation that concomitant L loa infection increases the risk of severe treatment-associated, life-threatening complications. Initiatives were therefore launched to map the risk of loiasis. Insight about the epidemiology of L loa has advanced notably; however, its effect on the individual as well as on the community level has not been well studied. In the absence of appropriate studies, L loa is commonly judged a harmless nematode, and loiasis as a separate entity does not belong to the list of neglected tropical diseases to be controlled or eradicated in worldwide campaigns. We advocate reorientation of research efforts towards a patient-centric view of loiasis and, as a first step, to establish the disease burden in disability-adjusted life-years of this chronic infection, and to answer the question of whether loiasis should be included in future control programmes.
Product Description
Supelco
Albendazole, analytical standard, ≥98%
USP
Albendazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Diethylcarbamazine citrate, European Pharmacopoeia (EP) Reference Standard
Albendazole, European Pharmacopoeia (EP) Reference Standard
Ivermectin, European Pharmacopoeia (EP) Reference Standard