Merck
  • Combination of fractional erbium-glass laser and topical therapy in melasma resistant to triple-combination cream.
Combination of fractional erbium-glass laser and topical therapy in melasma resistant to triple-combination cream.
The Journal of dermatological treatment (2012-03-06)
Athanasia Tourlaki, Michela Gianna Galimberti, Giovanni Pellacani, Pier Luca Bencini
ABSTRACT
Melasma is a common melanosis often difficult to treat. The aim of this paper was to report on the safety and efficacy of non-ablative fractional photothermolysis combined with the use of triple-combination cream (TCC) on a large population with melasma resistant (i.e., with no complete/near-complete clearing) to TCC alone. Seventy-six patients with resistant melasma underwent a combined treatment protocol. The protocol consisted of a TCC (hydroquinone 4%, retinoic acid 0.03%, hydrocortisone butyrate 0.1%) applied daily for 10 days followed by four laser treatments performed in 3-week intervals with a fractional 1540-nm erbium-glass laser. During these intervals, and for 3 months after the last laser session, TCC was also applied daily following a "pulse-therapy" scheme. Improvement was assessed by the melasma-area-and-severity-index (MASI) score. At 1 month, marked (>75%) and moderate (51-75%) clearing of melasma were observed in 46 of 76 (67.1%) and 12 of 76 (21%) cases, respectively. At 6 months, we noticed a marked improvement in 16 of 76 (21.1%) and no improvement in 33 of 76 (43.4%) patients. Our study proposes the combination of NFP/TCC as a useful therapy for patients with melasma resistant to TCC alone, but it shows that its long-term efficacy is limited.
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard