Merck
  • Formation of cyclic 1,N2-propanodeoxyguanosine adducts in DNA upon reaction with acrolein or crotonaldehyde.
Formation of cyclic 1,N2-propanodeoxyguanosine adducts in DNA upon reaction with acrolein or crotonaldehyde.
Cancer research (1984-03-01)
F L Chung, R Young, S S Hecht
ABSTRACT
Acrolein reacted with deoxyguanosine at pH 7 and 37 degrees to give three major products, Adducts 1 to 3, which were separated by high-performance liquid chromatography. They were identified by their ultraviolet, mass, and nuclear magnetic resonance spectra, by the spectra of the corresponding guanine derivatives, and by chemical transformations. Adducts 1 and 2 were two rapidly equilibrating diastereomers of 3-(2-deoxy-beta-D-erythro-pentofuranosyl)-5,6,7,8-tetrahydro-6- hydroxypyrimido [1,2-a]purine-10(3H)one, and Adduct 3 was 3-(2-deoxy-beta-D-erythro-pentofuranosyl)-5,6,7,8-tetrahydro-8- hydroxypyrimido [1,2-a]purine-10(3H)one. Adducts 1 and 2 were formed by Michael addition of N-1 of deoxyguanosine to C-3 of acrolein, followed by ring closure between N2 of deoxyguanosine and C-1 of acrolein. Adduct 3 was formed by ring closure in the opposite direction. Adduct 3 was analogous to the major crotonaldehyde-deoxyguanosine adducts which were previously characterized. Adduct 3 (0.2 mmol/mol DNA-P) or the corresponding crotonaldehyde adduct (0.03 mmol/mol DNA-P) was formed when either acrolein or crotonaldehyde was allowed to react with DNA at pH 7 and 37 degrees. These results demonstrate that cyclic 1,N2-propanodeoxyguanosine adducts are formed by reaction of acrolein and crotonaldehyde with DNA.
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer