Merck
  • Trace level analysis of corky off-flavor compounds: development of a new analytical method based on solid phase extraction and analysis by multidimensional gas chromatography with mass spectrometric detection.
Trace level analysis of corky off-flavor compounds: development of a new analytical method based on solid phase extraction and analysis by multidimensional gas chromatography with mass spectrometric detection.
Journal of chromatography. A (2011-11-08)
Hans-Georg Schmarr, Stefan Koschinski, Wei Sang, Petra Slabizki
ABSTRACT
This work describes the development of a trace level (<1 ngL(-1)) analysis of haloanisoles in complex wine matrix. The suggested method involves sample preparation based on solid phase extraction, a clean-up to remove acidic compounds, concentration of the haloanisole fraction and large volume on-column injection into a multidimensional GC-MS system. Mass spectrometric detection in the selected ion mode allowed reliable quantification of 2,4,6-trichloroanisole (TCA) or 2,4,6-tribromoanisole (TBA), via their highly deuterated ([(2)H5]) isotopologues as internal standards (stable isotope dilution analysis; SIDA), which had prior been synthesized in house. The development of this new method had become necessary, as a one-dimensional HS-SPME-GC-ECD method, routinely applied for analysis of TCA in cork soaks, had to be extended for TeCA and TBA determination, but failed due to co-elutions within wine matrices. The newly developed SPE//MDGC-MS method provided detection limits well below olfactory thresholds of the analytes with 0.05 ngL(-1) (LOD), 0.19 ngL(-1) (LOQ) for TCA, 0.06 ngL(-1) (LOD), 0.21 ngL(-1) (LOQ) for TeCA, and 0.09 ngL(-1) (LOD), 0.34 ngL(-1) (LOQ) for TBA.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler