Merck
  • Determination of benzenic and halogenated volatile organic compounds in animal-derived food products by one-dimensional and comprehensive two-dimensional gas chromatography-mass spectrometry.
Determination of benzenic and halogenated volatile organic compounds in animal-derived food products by one-dimensional and comprehensive two-dimensional gas chromatography-mass spectrometry.
Journal of chromatography. A (2009-09-29)
Jérémy Ratel, Erwan Engel
ABSTRACT
Animal-derived products are particularly vulnerable to contamination by volatile organic compounds (VOCs). These lipophilic substances, which are generated by an increasing number of sources, are easily transferred to the atmosphere, water, soil, and plants. They are ingested by livestock and become trapped in the fat fraction of edible animal tissues. The aim of this work was to determine the occurrence, risk for human health and entryways of benzenic and halogenated VOCs (BHVOCs) in meat products, milks and sea foods using gas chromatography- mass spectrometry (GC-MS) techniques. In the first part, the occurrence and levels of the BHVOCs in animal products were studied. One muscle and three fat tissues were analysed by GC-Quad/MS in 16 lambs. Of 52 BHVOCs identified, 46 were found in the three fat tissues and 29 in all four tissues, confirming that VOCs are widely disseminated in the body. Twenty-six BHVOCs were quantified in fat tissues, and risk for consumer health was assessed for six of these compounds regulated by the US Environmental Protection Agency (EPA). The BHVOC content was found to be consistent with previous reports and was below the maximum contaminant levels set by the EPA. In the second part, the performance of GCxGC-TOF/MS for comprehensively detecting BHVOCs and showing their entryways in animal-derived food chains was assessed. Meat, milk and oysters were analysed by GC-Quad/MS and GCxGC-TOF/MS. For all these products, at least a 7-fold increase in the contaminants detected was achieved with the GCxGC-TOF/MS technique. The results showed that the production surroundings, through animal feeding or geographical location, were key determinants of BHVOC composition in the animal products.
Product Description
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Pricing and availability is not currently available.
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.