Merck
  • Design of molecular photonic wires based on multistep electronic excitation transfer.
Design of molecular photonic wires based on multistep electronic excitation transfer.
Chemphyschem : a European journal of chemical physics and physical chemistry (2005-03-09)
Philip Tinnefeld, Mike Heilemann, Markus Sauer
ABSTRACT
Light-harvesting complexes, one of nature's supreme examples of nanoscale engineering, have inspired researchers to construct molecular optical devices, such as photonic wires, which are optimised for efficient transfer of excited-state energy over large distances. The control parameters for the design and the advantages of single-molecule fluorescence spectroscopy for the study of such complex systems are discussed with respect to energy-transfer mechanisms, chromophore selection and arrangement as well as static and dynamic heterogeneity.
Sigma-Aldrich
Atto 680 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)