- High-performance liquid chromatographic comparison of in vivo and in vitro drug metabolism.
High-performance liquid chromatographic comparison of in vivo and in vitro drug metabolism.
Journal of chromatography (1984-05-04)
W Voelter, T Kronbach
PMID6736160
ABSTRACT
Using 2-ethyl-3-(4- hydroxybenzoyl )benzofuran ( EHBB ) as an example, biotransformation in rabbits and rats and by rat hepatocyte suspensions was studied by high-performance liquid chromatography (HPLC) and mass spectrometry. The biotransformation of N-alkyl-substituted piperidines by rat hepatocytes gives valuable information about the pharmacodynamics of this series of compounds. It is demonstrated that a simple reversed-phase HPLC pre-column technique is much superior to the classical sample purification via extraction. The utility of hepatocytes for the investigation of drug metabolism is demonstrated.
Product Description
Benzarone, European Pharmacopoeia (EP) Reference Standard