Merck
Insulin sensitizers in polycystic ovary syndrome.
Frontiers of hormone research (2013-09-05)
Renato Pasquali, Alessandra Gambineri
ABSTRACT
From the conceptual point of view, there are several reasons to expect that improvement of insulin sensitivity may produce several benefits in the treatment of a complex disorder like polycystic ovary syndrome (PCOS), including a decrease in insulin and androgen levels, improvement of metabolic comorbidities, and, finally, improved ovulation and fertility. This can be achieved with the help of specific agents, particularly metformin and thiazolidinediones. They may ease the suffering of women with PCOS because insulin resistance and hyperinsulinemia appear to be major contributors to the pathophysiology of the syndrome.
Sigma-Aldrich
Enclomiphene hydrochloride, ≥96% (HPLC)