Merck
  • Effects of Leonurine on L-type calcium channel in rat ventricular myocytes.
Effects of Leonurine on L-type calcium channel in rat ventricular myocytes.
Biological & pharmaceutical bulletin (2012-08-07)
Hong Xin, Ming Gu, Wen Wei Wang, Shi Ying Huang, Fang Ping Li, Hui Cai, Yi Zhun Zhu, Xue Mei Zhang
ABSTRACT
Leonurine (Leo) is a special alkaloid principle of Herba leonuri that has recently been suggested to improve cardiovascular functions. To date, there is no direct ionic evidence of Leo on regulating calcium channels in the heart. In the present study, we examined the effects of Leo on action potentials and membrane currents recorded from isolated rat ventricular myocytes with the whole-cell patch clamp technique. Leo 100 µM shortened the action potential duration in a dose-dependent manner. Leo up to 200 µM had no significant effect on the Na+ current (INa) and K+ current (IK). However, Leo depressed the L-type Ca2+ current (ICa,L). In the presence of 20 and 100 µM Leo, the current density was decreased and the voltage at half maximal inactivation V0.5 shift to more negative potential. The recovery time constant was also delayed. In addition, the transcription and protein expression levels of L-type calcium channel (Cav1.2) in primary cultured neonatal myocytes from Sprague-Dawley rats were reduced by Leo treatment in a dose-dependent fashion as assessed by reverse transcription-polymerase chain reaction (RT-PCR) and Western blot assays. We conclude that Leo inhibits L-type calcium channels in cardiomyocytes.
Sigma-Aldrich
Leonurine, ≥98% (HPLC)