Merck
  • [Comparative study of drug efficacy and drug additives between generic drugs and original drugs].
[Comparative study of drug efficacy and drug additives between generic drugs and original drugs].
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (2007-12-07)
Hiromi Katoh, Michiko Yoshii, Koichiro Ozawa
ABSTRACT
In the present study, we tested three kinds of sleeping drugs, consisting mainly of triazolam, brotizolam, and flunitrazepam, to compare the drug efficacy of generic drugs with that of original drugs. After these drugs were administered orally to mice, drug efficacy was evaluated in terms of ambulation, onset time of sleep, and duration of sleep in the open field test. For all kinds of sleep-inducing drugs, the drug efficacy of most generic drugs is not necessarily equal to that of the original drug. The main reason for the difference appears to be due to differences in the rate of absorption of the main drug. Any other differences between an original drug and a generic drug are caused by drug additives, the crystal form of the main drug, the formulation, and so on. In this study, the formulation was not the reason for the differences because all of the drugs were pulverized in a mortar and had no special coating. The drug additives for all the drugs are listed and the drug efficacy compared. Unfortunately, the information was not sufficient to shed any light on the differences in drug efficacy. For effective drug therapy, more information on drug additives should be provided.