Merck
Octafluoronaphthalene-diphenylacetylene (1/1).
Acta crystallographica. Section C, Crystal structure communications (2001-07-10)
J C Collings, A S Batsanov, J A Howard, T B Marder
ABSTRACT
The structure of the title complex, C10F8*C14H10, comprises mixed stacks of alternating diphenylacetylene and octafluoronaphthalene molecules, both lying at inversion centres and parallel to within 8.6 (1) degrees, in contrast with the herring-bone packing observed in crystals of either pure component.
Sigma-Aldrich
Octafluoronaphthalene, 96%