Merck
  • Plasma LL-37 correlates with vitamin D and is reduced in human immunodeficiency virus-1 infected individuals not receiving antiretroviral therapy.
Plasma LL-37 correlates with vitamin D and is reduced in human immunodeficiency virus-1 infected individuals not receiving antiretroviral therapy.
Journal of medical microbiology (2014-05-14)
Jennifer R Honda, Elizabeth Connick, Samantha MaWhinney, Edward D Chan, Sonia C Flores
ABSTRACT
Low levels of the vitamin D-regulated antimicrobial peptide cathelicidin (LL-37) may negatively impact the immune status of human immunodeficiency virus-1 (HIV-1) infected individuals (HIV+). We compared plasma LL-37 levels in healthy controls (HIV-) and HIV+ individuals on or off antiretroviral therapies (ARTs) (ART+ and ART-, respectively), and evaluated the relationship between vitamin D and LL-37 levels. In this cross-sectional study, levels of LL-37, 25-hydroxycholecalciferol [25(OH)D3] and 1,25-dihydroxycholecalciferol [1,25(OH)2D3] were measured from an initial cohort of 18 healthy controls and 10 HIV+/ART- individuals. Because this cohort lacked HIV+/ART+ subjects, LL-37 was also quantified from a second cohort of 10 HIV+/ART- and 13 HIV+/ART+ individuals. LL-37 levels were significantly lower in the HIV+/ART- group compared to the healthy controls (P = 0.01). A direct relationship was observed between LL-37 and both 25(OH)D3 and 1,25(OH)2D3. The level of 25(OH)D3 was predictive of higher LL-37 (P = 0.04) and for any given level of 25(OH)D3, HIV+/ART- subjects averaged 20 % lower LL-37 compared to the healthy controls (P = 0.045). For any given level of 1,25(OH)2D3, HIV+/ART- subjects averaged 25% lower LL-37 compared to the healthy controls (P = 0.018), although 1,25(OH)2D3 was not predictive of higher LL-37 (P = 0.28). Finally, LL-37 levels were significantly lower in the HIV+/ART- group compared to the HIV+/ART+ group from the second cohort (P = 0.045). Untreated HIV infection may contribute to lower LL-37 levels, independent of vitamin D levels. ART treatment may potentially mitigate this decrease in LL-37 levels.
Product Description
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)