Merck
  • Syndecan binding protein (SDCBP) is overexpressed in estrogen receptor negative breast cancers, and is a potential promoter for tumor proliferation.
Syndecan binding protein (SDCBP) is overexpressed in estrogen receptor negative breast cancers, and is a potential promoter for tumor proliferation.
PloS one (2013-03-28)
Xiao-Long Qian, Ya-Qing Li, Bin Yu, Feng Gu, Fang-Fang Liu, Wei-Dong Li, Xin-Min Zhang, Li Fu
ABSTRACT
Syndecan binding protein (SDCBP), an adapter protein containing PDZ domains, contributes to the tumorigenicity and metastasis of many malignant tumors, such as malignant melanoma. Our study aimed in revealing the expression profile of SDCBP in breast cancer (BCa) and its role in tumor cell proliferation, and then exploring its value in the targeted treatment of BCa. We first evaluated the SDCBP expression by immunohistochemistry in normal breast and BCa tissues. Then we explored the expression profile of SDCBP in different BCa cell lines. By constructing SDCBP-silenced BCa cell clones, we further assessed the effects of SDCBP suppression on tumor cells in vitro by cell culture and in vivo by tumorigenicity. SDCBP expression was detected in 80.6% (n = 160) of BCa tissues, in contrast to its expression in 13% (n = 23) of normal breast tissues (P<0.001). Among the tumors, the level of its expression was positively correlated with histological grade and tumor staging while negatively correlated with the estrogen receptor (ER) expression. Higher expression of SDCBP was also noted in ER-negative BCa cell lines. It was also identified that SDCBP expression was down-regulated in a dose-dependent mode by 17-β estradiol in estrogen-responsive MCF-7. Furthermore, SDCBP silence inhibited ER-negative tumor cell growth in vivo and in vitro. Cell cycle studies showed that SDCBP silence increased G1 cell population and resulted in related cell-cycle-regulator changes: up-regulation of p21 and p27 while down-regulation of cyclin E. Our results suggested that SDCBP played an important role in tumor growth of ER-negative BCas. In these tumors where the estrogen signaling pathway is not available, SDCBP probably contribute to tumor growth through an alternative signaling pathway by promoting tumor cells passing the G1/S checkpoint into the cell cycle. Suppression of SDCBP expression may have its potential to become a targeted therapy for ER-negative BCas.
Sigma-Aldrich
Anti-SDCBP antibody produced in rabbit, IgG fraction of antiserum