Merck
  • Generation of transgenic mice with megabase-sized human yeast artificial chromosomes by yeast spheroplast-embryonic stem cell fusion.
Generation of transgenic mice with megabase-sized human yeast artificial chromosomes by yeast spheroplast-embryonic stem cell fusion.
Nature protocols (2013-07-23)
Liangping Li, Thomas Blankenstein
ABSTRACT
Introducing human genes into mice offers the opportunity to analyze their in vivo function or to obtain therapeutic molecules. For proper gene regulation, or in case of multigene families, megabase (Mb)-sized DNA fragments often have to be used. Yeast artificial chromosome (YAC)-mediated transgenesis is irreplaceable for this purpose, because alternative methods such as the use of bacterial artificial chromosomes (BACs) cannot introduce DNA fragments larger than 500 kb into the mouse germ line. However, YAC libraries often contain only partial gene loci. Time-consuming reconstruction of YACs, genetic instability and the difficulty in obtaining intact YAC DNA above a certain size impede the generation of humanized mice. Here we describe how to reconstruct YACs containing Mb-sized human DNA, such as the T cell receptor-α (TRA) gene locus, thus facilitating the introduction of large DNA fragments into the mouse germ line. Fusion of YAC-containing yeast and embryonic stem (ES) cells avoids the need for YAC DNA purification. These ES cells are then used to stably introduce the functional TRA gene locus into the mouse germ line. The protocol takes ∼1 year to complete, from reconstruction of the entire TRA gene locus from YACs containing partial but overlapping TRA regions to germline transmission of the YAC.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, 99.0-101.0% (titration)
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Roche
PCR DIG Probe Synthesis Kit, sufficient for 25 reaction (50 μL final reaction volume)