Merck
  • Long-term neuroprotective effects of NT-4-engineered mesenchymal stem cells injected intravitreally in a mouse model of acute retinal injury.
Long-term neuroprotective effects of NT-4-engineered mesenchymal stem cells injected intravitreally in a mouse model of acute retinal injury.
Investigative ophthalmology & visual science (2013-11-23)
Anna Machalińska, Miłosz Kawa, Ewa Pius-Sadowska, Jacek Stępniewski, Witold Nowak, Dorota Rogińska, Katarzyna Kaczyńska, Bartłomiej Baumert, Barbara Wiszniewska, Alicja Józkowicz, Józef Dulak, Bogusław Machaliński
ABSTRACT
Retinal degenerative diseases targeting the RPE and adjacent photoreceptors affect millions of people worldwide. The field of stem cell- and gene-based therapy holds great potential for the treatment of such diseases. The present study sought to graft genetically engineered mesenchymal stem cells (MSCs) that continuously produce neurotrophin-4 (NT-4) into the murine eye after the onset of acute retinal injury. C57BL/6 mice were subjected to acute retinal damage using a low dose of sodium iodate (20 mg/kg of body weight), followed by intravitreal injection of lentivirally modified MSC-NT-4 into the right eye. At 3 months after the MSC transplantation grafted cell survival, retinal function and gene expression were analyzed. Immunofluorescence analysis confirmed that transplanted MSCs survived for at least 3 months after intravitreal injection and preferentially migrated toward sites of injury within the retina. MSC-NT-4 actively produced NT-4 in the injured retina and significantly protected damaged retinal cells, as evaluated by ERG and optical coherence tomography (OCT). Of importance, the long-term therapy with MSC-NT-4 was also associated with induction of prosurvival signaling, considerable overexpression of some subsets of transcripts, including several members of the crystallin β-γ superfamily (Cryba4, Crybb3, Cryba2, Crybb1, Crybb2, Cryba1, and Crygc) and significant upregulation of biological processes associated with visual perception, sensory perception of light stimulus, eye development, sensory organ development, and system development. Transplantation of genetically modified MSCs that produce neurotrophic growth factors may represent a useful strategy for treatment of different forms of retinopathies in the future.
Sigma-Aldrich
Sodium iodate, puriss. p.a., ≥99.5% (RT)
Sigma-Aldrich
Sodium iodate, ≥99%