Merck
  • Obstructive sleep apnea syndrome affects liver histology and inflammatory cell activation in pediatric nonalcoholic fatty liver disease, regardless of obesity/insulin resistance.
Obstructive sleep apnea syndrome affects liver histology and inflammatory cell activation in pediatric nonalcoholic fatty liver disease, regardless of obesity/insulin resistance.
American journal of respiratory and critical care medicine (2013-11-22)
Valerio Nobili, Renato Cutrera, Daniela Liccardo, Martino Pavone, Rita Devito, Valentina Giorgio, Elisabetta Verrillo, Giuseppe Baviera, Giovanni Musso
ABSTRACT
Obstructive sleep apnea syndrome (OSAS) and nonalcoholic fatty liver disease (NAFLD) are frequently encountered in obese children. Whether OSAS and intermittent hypoxia are associated with liver injury in pediatric NAFLD is unknown. To assess the relationship of OSAS with liver injury in pediatric NAFLD. Sixty-five consecutive children with biopsy-proven NAFLD (age, mean ± SD, 11.7 ± 2.1 yr; 58% boys; body mass index z score, 1.93 ± 0.61) underwent a clinical-biochemical assessment and a standard polysomnography. Insulin sensitivity, circulating proinflammatory cytokines, markers of hepatocyte apoptosis (cytokeratin-18 fragments), and hepatic fibrogenesis (hyaluronic acid) were measured. Liver inflammatory infiltrate was characterized by immunohistochemistry for CD45, CD3, and CD163, surface markers of leukocytes, T cells, and activated macrophage/Kupffer cells, respectively. OSAS was defined by an apnea/hypopnea index (AHI) greater than or equal to 1 event/h, and severe OSAS was defined by an AHI greater than or equal to 5 events/h. Fifty-five percent of children with NAFLD had nonalcoholic steatohepatitis (NASH), and 34% had significant (stage F ≥ 2) fibrosis. OSAS affected 60% of children with NAFLD; the presence and severity of OSAS were associated with the presence of NASH (odds ratio, 4.89; 95% confidence interval, 3.08-5.98; P = 0.0001), significant fibrosis (odds ratio, 5.91; 95% confidence interval, 3.23-7.42; P = 0.0001), and NAFLD activity score (β, 0.347; P = 0.029), independently of body mass index, abdominal adiposity, metabolic syndrome, and insulin resistance. This relationship held also in nonobese children with NAFLD. The duration of hemoglobin desaturation (Sa(O2) < 90%) correlated with increased intrahepatic leukocytes and activated macrophages/Kupffer cells and with circulating markers of hepatocyte apoptosis and fibrogenesis. In pediatric NAFLD, OSAS is associated with biochemical, immunohistochemical, and histological features of NASH and fibrosis. The impact of hypoxemia correction on liver disease severity warrants evaluation in future trials.
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Sodium hyaluronate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000