Merck
  • Impedance spectroscopy of highly ordered nano-porous electrodes based on Au-AAO (anodic aluminum oxide) structure.
Impedance spectroscopy of highly ordered nano-porous electrodes based on Au-AAO (anodic aluminum oxide) structure.
Journal of nanoscience and nanotechnology (2013-11-20)
Jaehwan Ahn, Sungbo Cho, Junhong Min
ABSTRACT
Electrochemical measurements using the microelectrodes are increasingly utilized for the label-free detection of the small amount of biological materials such as DNA, protein, and cells. However, the interfacial electrode impedance increases and may hinder the detection of weak signals as the size of electrode decreases. To enhance the measurement sensitivity while reducing the electrode size, in this study, microelectrodes employing a nanoporous structure were fabricated and characterized by using electrical impedance spectroscopy. We made the highly ordered honeycomb nanoporous structure of Anodic Aluminum Oxide (AAO) by electrochemical anodizing and formed Au layer on the surface of AAO (Au/AAO) by electroless Au plating method. The electrical characteristics of the fabricated Au/AAO electrodes were evaluated by using de Levie's model derived for the pore electrodes. As a result, the interfacial electrode impedance of the fabricated Au/AAO electrodes was 2-3 order lower than the value of the planar electrodes at frequencies below 1 kHz. It implies this nanoporous electrode could be directly applied to label free detection of biomaterials.
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh