Merck
  • Metabolic flux between unsaturated and saturated fatty acids is controlled by the FabA:FabB ratio in the fully reconstituted fatty acid biosynthetic pathway of Escherichia coli.
Metabolic flux between unsaturated and saturated fatty acids is controlled by the FabA:FabB ratio in the fully reconstituted fatty acid biosynthetic pathway of Escherichia coli.
Biochemistry (2013-10-24)
Xirui Xiao, Xingye Yu, Chaitan Khosla
ABSTRACT
The entire fatty acid biosynthetic pathway of Escherichia coli, starting from the acetyl-CoA carboxylase, has been reconstituted in vitro from 14 purified protein components. Radiotracer analysis verified stoichiometric conversion of acetyl-CoA and NAD(P)H to the free fatty acid product, allowing implementation of a facile spectrophotometric assay for kinetic analysis of this multienzyme system. At steady state, a maximal turnover rate of 0.5 s(-1) was achieved. Under optimal turnover conditions, the predominant products were C16 and C18 saturated as well as monounsaturated fatty acids. The reconstituted system allowed us to quantitatively interrogate the factors that influence metabolic flux toward unsaturated versus saturated fatty acids. In particular, the concentrations of the dehydratase FabA and the β-ketoacyl synthase FabB were found to be crucial for controlling this property. Via changes in these variables, the percentage of unsaturated fatty acid produced could be adjusted between 10 and 50% without significantly affecting the maximal turnover rate of the pathway. Our reconstituted system provides a powerful tool for understanding and engineering rate-limiting and regulatory steps in this complex and practically significant metabolic pathway.
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Acetyl-CoA carboxylase 2 human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Hydroquinidine, 95%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Acetyl-CoA Carboxylase 1 human, recombinant, expressed in Sf9 cells