Merck
  • Arabidopsis thaliana AMY3 is a unique redox-regulated chloroplastic α-amylase.
Arabidopsis thaliana AMY3 is a unique redox-regulated chloroplastic α-amylase.
The Journal of biological chemistry (2013-10-04)
David Seung, Matthias Thalmann, Francesca Sparla, Maher Abou Hachem, Sang Kyu Lee, Emmanuelle Issakidis-Bourguet, Birte Svensson, Samuel C Zeeman, Diana Santelia
ABSTRACT
α-Amylases are glucan hydrolases that cleave α-1,4-glucosidic bonds in starch. In vascular plants, α-amylases can be classified into three subfamilies. Arabidopsis has one member of each subfamily. Among them, only AtAMY3 is localized in the chloroplast. We expressed and purified AtAMY3 from Escherichia coli and carried out a biochemical characterization of the protein to find factors that regulate its activity. Recombinant AtAMY3 was active toward both insoluble starch granules and soluble substrates, with a strong preference for β-limit dextrin over amylopectin. Activity was shown to be dependent on a conserved aspartic acid residue (Asp(666)), identified as the catalytic nucleophile in other plant α-amylases such as the barley AMY1. AtAMY3 released small linear and branched glucans from Arabidopsis starch granules, and the proportion of branched glucans increased after the predigestion of starch with a β-amylase. Optimal rates of starch digestion in vitro was achieved when both AtAMY3 and β-amylase activities were present, suggesting that the two enzymes work synergistically at the granule surface. We also found that AtAMY3 has unique properties among other characterized plant α-amylases, with a pH optimum of 7.5-8, appropriate for activity in the chloroplast stroma. AtAMY3 is also redox-regulated, and the inactive oxidized form of AtAMY3 could be reactivated by reduced thioredoxins. Site-directed mutagenesis combined with mass spectrometry analysis showed that a disulfide bridge between Cys(499) and Cys(587) is central to this regulation. This work provides new insights into how α-amylase activity may be regulated in the chloroplast.
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.