Merck
  • Parallel analysis of volatile fatty acids, indole, skatole, phenol, and trimethylamine from waste-related source environments.
Parallel analysis of volatile fatty acids, indole, skatole, phenol, and trimethylamine from waste-related source environments.
Journal of chromatography. A (2013-09-28)
Md Mahmudur Rahman, Ki-Hyun Kim
ABSTRACT
An experimental technique based on sorbent tube-thermal desorption-gas chromatography (ST-TD-GC) was investigated for the simultaneous determination of a cluster of eight volatile odorants (propionic acid, n-butyric acid, i-valeric acid, n-valeric acid, trimethylamine, phenol, indole, and skatole) and a reference compound (benzene). Calibration was made by direct injection of a liquid working standard (L-WS) into a quartz tube packed with three bed sorbent (Tenax TA, Carbopack B, and Carbopack X). To assess the relative performance between different detector systems, a comparative analysis was made using both mass spectrometry (MS) and a flame ionization detector (FID) with the aid of a TD system. In the TD-GC-MS analysis, calibration results were evaluated in two different modes, namely total ion chromatogram (TIC) and extracted ion chromatogram (EIC). In both FID and MS, the elution order of investigated odorants complied with the retention time index (RTI) values for the polar column with a coefficient of determination (R(2)) at or above 0.99. As a means to validate our detection approach, environmental samples from a bathroom and manhole (vacuum samples) as well as cat stool and wastewater (headspace samples) were also collected. The ST-TD method tested for the concurrent analysis of diverse odorants allowed us to measure a list of offensive odorants from those samples.
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Sigma-Aldrich
Skatole, ≥98%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Trimethylamine hydrochloride, 98%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)