Merck
  • Spectrophotometeric determination of trace amounts of Al3+ ion in water samples after cloud point extraction using quinizarin as a complexing agent.
Spectrophotometeric determination of trace amounts of Al3+ ion in water samples after cloud point extraction using quinizarin as a complexing agent.
Environmental monitoring and assessment (2013-09-26)
Ardeshir Shokrollahi, Roghayeh Aghaei
ABSTRACT
In this study, cloud point extraction was used for the preconcentration of Al(3+) ion after the complex formation with 1,4-dihydroxy-9,10-anthraquinone (Quinizarin [QUIN]), and subsequent analysis by spectrophotometeric method, using Triton X-114 as surfactant. The optimal extraction and reaction conditions were studied (i.e., pH = 5.5, 0.1 mM QUIN, Triton X-114 = 0.1% (w/v)), and the analytical characteristics of the method (e.g., limit of detection, linear range, preconcentration, and enrichment factors) were obtained. Linearity was obeyed in the range of 3.33-166.67 ng ml(-1) of Al(3+) ion. The detection limit of the method was 2.09 ng ml(-1) for Al(3+) ion. The interference effect of some anions and cations was also tested. The method was applied to determine Al(3+) ion in water samples.
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis