Merck
  • Pre-storage application of oxalic acid alleviates chilling injury in mango fruit by modulating proline metabolism and energy status under chilling stress.
Pre-storage application of oxalic acid alleviates chilling injury in mango fruit by modulating proline metabolism and energy status under chilling stress.
Food chemistry (2013-09-05)
Peiyan Li, Xiaolin Zheng, Yan Liu, Yuyan Zhu
ABSTRACT
Effects of oxalic acid on chilling injury, proline metabolism and energy status in mango fruit were investigated after mango fruit (Mangifera indica L. cv. Zill) were dipped in 5mM oxalic acid solution for 10min at 25°C and then stored at low temperature (10±0.5°C) for 49days thereafter transferred to 25°C for 4days. Pre-storage application of oxalic acid apparently inhibited the development of chilling injury, notably elevated proline accumulation actually associated with increase in Δ(1)-pyrroline-5-carboxylate synthetase (P5CS) activity and decrease in proline dehydrogenase (PDH) activity in the peel and the flesh, without activation of ornithine-δ-aminotransferase (OAT) activity, and maintained high ATP level and energy charge in the flesh during storage. It was suggested that these effects of oxalic acid might collectively contribute to improving chilling tolerance, thereby alleviating chilling injury and maintaining quality of mango fruit in long term cold storage.
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis