Merck
  • Exploratory use of cardiovascular magnetic resonance imaging in liver transplantation: a one-stop shop for preoperative cardiohepatic evaluation.
Exploratory use of cardiovascular magnetic resonance imaging in liver transplantation: a one-stop shop for preoperative cardiohepatic evaluation.
Transplantation (2013-08-29)
Sahadev T Reddy, Ngoc L Thai, Asghar A Fakhri, Jose Oliva, Kusum B Tom, Michael K Dishart, Mark Doyle, June A Yamrozik, Ronald B Williams, Saundra B Grant, Jacqueline Poydence, Moneal Shah, Anil Singh, Swami Nathan, Robert W W Biederman
ABSTRACT
Preoperative cardiovascular risk stratification in orthotopic liver transplantation candidates has proven challenging due to limitations of current noninvasive modalities. Additionally, the preoperative workup is logistically cumbersome and expensive given the need for separate cardiac, vascular, and abdominal imaging. We evaluated the feasibility of a "one-stop shop" in a magnetic resonance suite, performing assessment of cardiac structure, function, and viability, along with simultaneous evaluation of thoracoabdominal vasculature and liver anatomy. In this pilot study, patients underwent steady-state free precession sequences and stress cardiac magnetic resonance (CMR), thoracoabdominal magnetic resonance angiography, and abdominal magnetic resonance imaging (MRI) on a standard MRI scanner. Pharmacologic stress was performed using regadenoson, adenosine, or dobutamine. Viability was assessed using late gadolinium enhancement. Over 2 years, 51 of 77 liver transplant candidates (mean age, 56 years; 35% female; mean Model for End-stage Liver Disease score, 10.8; range, 6-40) underwent MRI. All referred patients completed standard dynamic CMR, 98% completed stress CMR, 82% completed late gadolinium enhancement for viability, 94% completed liver MRI, and 88% completed magnetic resonance angiography. The mean duration of the entire study was 72 min, and 45 patients were able to complete the entire examination. Among all 51 patients, 4 required follow-up coronary angiography (3 for evidence of ischemia on perfusion CMR and 1 for postoperative ischemia), and none had flow-limiting coronary disease. Nine proceeded to orthotopic liver transplantation (mean 74 days to transplantation after MRI). There were six ascertained mortalities in the nontransplant group and one death in the transplanted group. Explant pathology confirmed 100% detection/exclusion of hepatocellular carcinoma. No complications during CMR examination were encountered. In this proof-of-concept study, it appears feasible to perform a comprehensive, efficient, and safe preoperative liver transplant imaging in a CMR suite-a one-stop shop, even in seriously ill patients.
Sigma-Aldrich
Dobutamine hydrochloride, ≥98%
Dobutamine hydrochloride, European Pharmacopoeia (EP) Reference Standard