Merck
  • Comparison of poliglecaprone-25 and polyglactin-910 in cutaneous surgery.
Comparison of poliglecaprone-25 and polyglactin-910 in cutaneous surgery.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.] (2013-08-15)
Thomas Regan, Naomi Lawrence
ABSTRACT
Few clinical studies have compared deep absorbable sutures. Poliglecaprone-25 and polyglactin-910 are two of the most commonly used absorbable sutures in cutaneous surgery. To compare the rate of suture extrusion, degree of lumpiness, and appearance of scars from wounds closed with poliglecaprone-25 and polyglactin-910. Poliglecaprone-25 or polyglactin-910 was used for closure of the deep part of Mohs defects. The number of extruded sutures and the number of lumps were recorded at each follow-up visit. Photographs of 1-week and 3-month postoperative scars were rated on a visual analogue scale. One hundred forty patients completed the study. There was a statistically significant difference in the percentage of extruded sutures between poliglecaprone-25 (3.1%) and polyglactin-910 (11.4%) (p < .01). There was not a statistically significant difference in the percentage of lumps (both 22%) or overall appearance of scars at 1 week or 3 months. Poliglecaprone-25 resulted in significantly less extruded sutures than did polyglactin-910, although both resulted in the same degree of lumpiness and similar-appearing scars at 1 week and 3 months.
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000