Merck
Molecular rotors of coronene in charge-transfer solids.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-07-31)
Yukihiro Yoshida, Yasuhiro Shimizu, Takeshi Yajima, Goro Maruta, Sadamu Takeda, Yoshiaki Nakano, Takaaki Hiramatsu, Hiroshi Kageyama, Hideki Yamochi, Gunzi Saito
ABSTRACT
Ten types of neutral charge transfer (CT) complexes of coronene (electron donor; D) were obtained with various electron acceptors (A). In addition to the reported 7,7,8,8-tetracyanoquinodimethane (TCNQ) complex of 1:1 stoichiometry with a DA-type alternating π column, TCNQ also afforded a 3:1 complex, in which a face-to-face dimer of parallel coronenes (Cor-As) is sandwiched between TCNQs to construct a DDA-type alternating π column flanked by another coronene (Cor-B). Whereas solid-state (2)H NMR spectra of the 1:1 TCNQ complex formed with deuterated coronene confirmed the single in-plane 6-fold flipping motion of the coronenes, two unsynchronized motions were confirmed for the 3:1 TCNQ complex, which is consistent with a crystallographic study. Neutral [Ni(mnt)2] (mnt: maleonitriledithiolate) as an electron acceptor afforded a 5:2 complex with a DDA-type alternating π column flanked by another coronene, similar to the 3:1 TCNQ complex. The fact that the Cor-As in the [Ni(mnt)2] complex arrange in a non-parallel fashion must cause the fast in-plane rotation of Cor-A relative to that of Cor-B. This is in sharp contrast to the 3:1 TCNQ complex, in which the dimer of parallel Cor-As shows inter-column interactions with neighboring Cor-As. The solid-state (1)H NMR signal of the [Ni(mnt)2] complex suddenly broadens at temperatures below approximately 60 K, indicating that the in-plane rotation of the coronenes undergoes down to approximately 60 K; the rotational rate reaches the gigahertz regime at room temperature. Rotational barriers of these CT complexes, as estimated from variable-temperature spin-lattice relaxation time (T1) experiments, are significantly lower than that of pristine coronene. The investigated structure-property relationships indicate that the complexation not only facilitates the molecular rotation of coronenes but also provides a new solid-state rotor system that involves unsynchronized plural rotators.
Sigma-Aldrich
7,7,8,8-Tetracyanoquinodimethane, 98%
Coronene, BCR®, certified reference material
Sigma-Aldrich
Coronene, 97%
Sigma-Aldrich
Coronene, purified by sublimation, 99%