Merck
Inhibition of human platelet aggregation by eosinophils.
Life sciences (2013-07-23)
Aline Mendes Maziero, Raquel Lorenzetti, José Luiz Donato, Sergio Lilla, Gilberto De Nucci
ABSTRACT
The relationship between the activity of eosinophils and platelets has been observed in recent decades by many scientists. These observations include increased numbers of eosinophils associated with platelet disorders, including changes in the coagulation cascade and platelet aggregation. Based on these observations, the interaction between eosinophils and platelets in platelet aggregation was analyze. Human platelets were incubated with eosinophil cytosolic fraction, promyelocytic human HL-60 clone 15 cell lineage, and eosinophil cationic protein (ECP). Platelet rich plasma (PRP) aggregation was induced by adenosine diphosphate, platelet activating factor, arachidonic acid, and collagen, and washed platelets (WP) were activated by thrombin. Aggregation induced by all agonists was dose dependently inhibited by eosinophil cytosolic fraction. This inhibition was only partially reversed by previous incubation of the eosinophils with l-Nitro-Arginine-Methyl-Ester (l-NAME). Previous incubation with indomethacin did not prevent the cytosolic fraction induced inhibition. The separation of eosinophil cytosolic fraction by gel filtration on Sephadex G-75 showed that the inhibitory activity was concentrated in the lower molecular weight fraction. HL-60 clone 15 cells differentiated into eosinophils for 5 and 7 day were able to inhibit platelet aggregation. The ECP protein inhibited the platelet aggregation on PRP and WP. This inhibition was more evident in WP, and the citotoxicity MTT assay proved the viability of tested platelets, showing that the observed inhibition by the ECP protein does not occur simply by cell death. Our results indicate that eosinophils play a fundamental role in platelet aggregation inhibition.
Product Description
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran, analytical standard, for GPC, 670,000
Supelco
Dextran, analytical standard, for GPC, 150,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, 5,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Sephadex® G-100, BioReagent, for molecular biology, DNA grade, medium
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Sephadex® G-25, BioReagent, for molecular biology, DNA grade, Superfine
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)