Merck
  • Inherently electroactive graphene oxide nanoplatelets as labels for specific protein-target recognition.
Inherently electroactive graphene oxide nanoplatelets as labels for specific protein-target recognition.
Nanoscale (2013-07-13)
Adeline Huiling Loo, Alessandra Bonanni, Martin Pumera
ABSTRACT
Graphene related materials have been widely employed as highly efficient transducers for biorecognition. Here we show a conceptually new approach of using graphene oxide nanoplatelets (50 × 50 nm) as voltammetric inherently active labels for specific protein-target molecule recognition. This proof-of-principle is demonstrated by biotin-avidin recognition, which displays that graphene oxide nanoplatelet labels show excellent selectivity. Therefore, it is expected that inherently electroactive graphene oxide nanoplatelet labels will play a similar role as electroactive gold nanoparticle labels which were developed more than a decade ago.
Product Description
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%