Merck
  • Decreased long-chain fatty acid oxidation impairs postischemic recovery of the insulin-resistant rat heart.
Decreased long-chain fatty acid oxidation impairs postischemic recovery of the insulin-resistant rat heart.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-07-05)
Romain Harmancey, Hernan G Vasquez, Patrick H Guthrie, Heinrich Taegtmeyer
ABSTRACT
Diabetic patients with acute myocardial infarction are more likely to die than nondiabetic patients. In the present study we examined the effect of insulin resistance on myocardial ischemia tolerance. Hearts of rats, rendered insulin resistant by high-sucrose feeding, were subjected to ischemia/reperfusion ex vivo. Cardiac power of control hearts from chow-fed rats recovered to 93%, while insulin-resistant hearts recovered only to 80% (P<0.001 vs. control). Unexpectedly, impaired contractile recovery did not result from an impairment of glucose oxidation (576±36 vs. 593±42 nmol/min/g dry weight; not significant), but from a failure to increase and to sustain oxidation of the long-chain fatty acid oleate on reperfusion (1878±56 vs. 2070±67 nmol/min/g dry weight; P<0.05). This phenomenon was due to a reduced ability to transport oleate into mitochondria and associated with a 38-58% decrease in the mitochondrial uncoupling protein 3 (UCP3) levels. Contractile function was rescued by replacing oleate with a medium-chain fatty acid or by restoring UCP3 levels with 24 h of food withdrawal. Lastly, the knockdown of UCP3 in rat L6 myocytes also decreased oleate oxidation by 13-18% following ischemia. Together the results expose UCP3 as a critical regulator of long-chain fatty acid oxidation in the stressed heart postischemia and identify octanoate as an intervention by which myocardial metabolism can be manipulated to improve function of the insulin-resistant heart.
Product Description
Supelco
Caprylic Acid (Octanoic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Caprylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Octanoic acid, ≥99%
Supelco
Octanoic acid, analytical standard
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG