Merck
  • Amperometric detection of dopamine in human serum by electrochemical sensor based on gold nanoparticles doped molecularly imprinted polymers.
Amperometric detection of dopamine in human serum by electrochemical sensor based on gold nanoparticles doped molecularly imprinted polymers.
Biosensors & bioelectronics (2013-06-12)
Cheng Xue, Qing Han, Yang Wang, Jinhua Wu, Tingting Wen, Ruoyu Wang, Junli Hong, Xuemin Zhou, Huijun Jiang
ABSTRACT
In this work, a highly sensitive and selective biomimetic electrochemical sensor for the amperometric detection of trace dopamine (DA) in human serums was achieved by gold nanoparticles (AuNPs) doped molecularly imprinted polymers (MIPs). Functionalized AuNPs (F-AuNPs), a novel functional monomer bearing aniline moieties on the surface of the AuNPs, were prepared via a direct synthesis method and then used to fabricate the conductive MIPs film on the modified electrode by electropolymerization method in the presence of DA and p-aminobenzenethiol (p-ATP). The obtained electrochemical sensor based on the conductive film of AuNPs doped MIPs (AuNPs@MIPs) could effectively minimize the interferences caused by ascorbic acid (AA) and uric acid (UA). The linear range for amperometric detection of DA was from 0.02 μmol L(-1) to 0.54 μmol L(-1) with the detection limit of 7.8 nmol L(-1) (S/N=3). Furthermore, the AuNPs@MIPs modified electrode (AuNPs@MIES) was successfully employed to detect trace DA in different human serums.
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline hydrochloride, ≥99%