Merck
  • Reduced renal clearance of cefotaxime in asians with a low-frequency polymorphism of OAT3 (SLC22A8).
Reduced renal clearance of cefotaxime in asians with a low-frequency polymorphism of OAT3 (SLC22A8).
Journal of pharmaceutical sciences (2013-05-08)
Sook Wah Yee, Anh Nguyet Nguyen, Chaline Brown, Radojka M Savic, Youcai Zhang, Richard A Castro, Cheryl D Cropp, Ji Ha Choi, Diment Singh, Harunobu Tahara, Sophie L Stocker, Yong Huang, Claire M Brett, Kathleen M Giacomini
ABSTRACT
Organic anion transporter 3 (OAT3, SLC22A8), a transporter expressed on the basolateral membrane of the proximal tubule, plays a critical role in the renal excretion of organic anions including many therapeutic drugs. The goal of this study was to evaluate the in vivo effects of the OAT3-Ile305Phe variant (rs11568482), present at 3.5% allele frequency in Asians, on drug disposition with a focus on cefotaxime, a cephalosporin antibiotic. In HEK293-Flp-In cells, the OAT3-Ile305Phe variant had a lower maximum cefotaxime transport activity, Vmax , [159 ± 3 nmol*(mg protein)(-1) /min (mean ± SD)] compared with the reference OAT3 [305 ± 28 nmol*(mg protein)(-1) /min, (mean ± SD), p < 0.01], whereas the Michaelis-Menten constant values (Km ) did not differ. In healthy volunteers, we found volunteers that were heterozygous for the Ile305Phe variant and had a significantly lower cefotaxime renal clearance (CLR ; mean ± SD: 84.8 ± 32.1 mL/min, n = 5) compared with volunteers that were homozygous for the reference allele (158 ± 44.1 mL/min, n = 10; p = 0.006). Furthermore, the net secretory component of cefotaxime renal clearance (CLsec ) was reduced in volunteers heterozygous for the variant allele [33.3 ± 31.8 mL/min (mean ± SD)] compared with volunteers homozygous for the OAT3 reference allele [97.0 ± 42.2 mL/min (mean ± SD), p = 0.01]. In summary, our study suggests that a low-frequency reduced-function polymorphism of OAT3 associates with reduced cefotaxime CLR and CL(sec) .
Sigma-Aldrich
Cefotaxime sodium salt, suitable for plant cell culture, BioReagent, powder or crystals
Sigma-Aldrich
Cefotaxime sodium salt, potency: 916-964 μg per mg
Cefotaxime acid, European Pharmacopoeia (EP) Reference Standard
Cefotaxime sodium, European Pharmacopoeia (EP) Reference Standard